Learn More
Ranitidine hydrochloride, 99%, Thermo Scientific™
£32.00 - £114.00
Chemical Identifiers
| CAS | 66357-59-3 |
|---|---|
| Molecular Formula | C13H23ClN4O3S |
| Molecular Weight (g/mol) | 350.862 |
| MDL Number | MFCD00069339 |
| InChI Key | GGWBHVILAJZWKJ-CHHCPSLASA-N |
| Synonym | ranitidine hydrochloride, noctone, n-2-5-dimethylamino methyl-2-furanyl methyl thio ethyl-n'-methyl-2-nitro-1,1-ethanediamine hydrochloride, c13h22n4o3s.hcl, opera_id_624, melfax hydrochloride, zantac hydrochloride, zintac hydrochloride, azantac hydrochloride, raniben hydrochloride |
| PubChem CID | 6603542 |
| IUPAC Name | (Z)-1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride |
| SMILES | CNC(=C[N+](=O)[O-])NCCSCC1=CC=C(O1)CN(C)C.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11495906
|
Thermo Scientific Alfa Aesar
B22260.06 |
5g |
£32.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11405916
|
Thermo Scientific Alfa Aesar
B22260.14 |
25g |
£114.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsAn H-2 histamine receptor antagonistRanitidine hydrochloride is mainly used as an H2-receptor antagonist and helps in the treatment of gastrointestinal lesions because of excessive gastric acid secretion. It also serves as an antiulcer drug.
Solubility
Soluble in water, 2-hydroxypropyl-beta-cyclodextrin, acetic acid, methanol, ethanol and dimethyl sulfoxide. Insoluble in chloroform.
Notes
Light sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
| 66357-59-3 | |
| 350.862 | |
| GGWBHVILAJZWKJ-CHHCPSLASA-N | |
| 6603542 | |
| CNC(=C[N+](=O)[O-])NCCSCC1=CC=C(O1)CN(C)C.Cl |
| C13H23ClN4O3S | |
| MFCD00069339 | |
| ranitidine hydrochloride, noctone, n-2-5-dimethylamino methyl-2-furanyl methyl thio ethyl-n'-methyl-2-nitro-1,1-ethanediamine hydrochloride, c13h22n4o3s.hcl, opera_id_624, melfax hydrochloride, zantac hydrochloride, zintac hydrochloride, azantac hydrochloride, raniben hydrochloride | |
| (Z)-1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride |
Specifications
| 66357-59-3 | |
| C13H23ClN4O3S | |
| 14,8110 | |
| Soluble in water,2-hydroxypropyl-beta-cyclodextrin,acetic acid,methanol,ethanol and dimethyl sulfoxide. Insoluble in chloroform. | |
| CNC(=C[N+](=O)[O-])NCCSCC1=CC=C(O1)CN(C)C.Cl | |
| 350.862 | |
| 350.86 | |
| Ranitidine hydrochloride |
| 99% | |
| MFCD00069339 | |
| ranitidine hydrochloride, noctone, n-2-5-dimethylamino methyl-2-furanyl methyl thio ethyl-n'-methyl-2-nitro-1,1-ethanediamine hydrochloride, c13h22n4o3s.hcl, opera_id_624, melfax hydrochloride, zantac hydrochloride, zintac hydrochloride, azantac hydrochloride, raniben hydrochloride | |
| GGWBHVILAJZWKJ-CHHCPSLASA-N | |
| (Z)-1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride | |
| 6603542 | |
| 99% |
Safety and Handling
missing translation for 'einecsNumber' : 266-333-0
missing translation for 'tsca' : No
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.