missing translation for 'onlineSavingsMsg'
Learn More

Ranitidine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30464252
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464252 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464252 Supplier LoGiCal Supplier No. LGCAMP0086.0001
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ranitidine Hydrochloride
CAS 66357-59-3
Analyte or Component Names Ranitidine Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C13 H22 N4 O3 S . Cl H
Certifications/Compliance ISO 17034
Synonym Ranitidine Hydrochloride, 1,1-Ethenediamine, N'-[2-[[[5-[(dimethylamino)methyl]-2-furanyl]methyl]thio]ethyl]-N-methyl-2-nitro-, hydrochloride (1:1), 1,1-Ethenediamine, N-[2-[[[5-[(dimethylamino)methyl]-2-furanyl]methyl]thio]ethyl]-N'-methyl-2-nitro-, monohydrochloride (9CI), AH 19065, Achedos, Acidex, Aciloc, Acloral, Aldin, Anistal, Antagonin, Apo-Ranitidine, Apozan, Atural, Axoban, Azantac, Consec, Coralen, Curan, Eltidine, Ezopta, Galidrin, Gastrial, Gastridin, Gastrosedol, Histac, Histak, Lydin, Melfax, Microtid, Neoceptin R, Noctone, Normon, Ptinolin, Quantor, Quicran, RND, Radinat, Ranacid, Randin, Rani 2, Ranial, Raniben, Ranidil, Ranidine, Ranigast, Ranimex, Ranin, Raniogas, Raniplex, Ranisen, Ranitab, Ranital, Ranitidine hydrochloride, Ranitidine-akri, Ranitiget, Ranitin, Ranix, Rantac, Rantacid, Rantidol, Rantin, Ranuber, Ratic, Raticina, Rosimol, Simetac, Sostril, Taural, Terposen, Trigger, Ul-Pep, Ulceran, Ulceranin, Ulcex, Ulcirex, Ulsal, Ultac, Ultidine, Urantac, Verlost, Vesyca, Vizerul, Weichilin, Weidos, Xanidine, Zantab, Zantac, Zantadin, Zantic, Zinetac, Zintac, Ranitidine hydrochloride (AH19065), AH 19065AB
InChI Formula InChI=1S/C13H22N4O3S.ClH/c1-14-13(9-17(18)19)15-6-7-21-10-12-5-4-11(20-12)8-16(2)3;/h4-5,9,14-15H,6-8,10H2,1-3H3;1H
Solution Type Single Solution
SMILES Cl.CNC(=C[N+](=O)[O-])NCCSCc1oc(CN(C)C)cc1
IUPAC Name 1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 350.86
Quantity 1 mL
Formula Weight 350.1179 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.