missing translation for 'onlineSavingsMsg'
Learn More

Ranitidine EP Impurity D (as Sodium Salt), Mikromol™

Product Code. 30050817
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050817 25 mg 25mg
17963046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050817 Supplier Mikromol Supplier No. MM0086.060025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ranitidine EP Impurity D (as Sodium Salt)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-[((5-[(Dimethylamino)methyl]furan-2-yl)-methyl)sulphanyl]ethyl]-2-nitroacetamide Sodium Salt
CAS 112251-56-6
Analyte or Component Names N-[2-[((5-[(Dimethylamino)methyl]furan-2-yl)-methyl)sulphanyl]ethyl]-2-nitroacetamide Sodium Salt
Shipping Condition Ice pack (-20°C)
Molecular Formula C12 H18 N3 O4 S . Na
Certifications/Compliance ISO 9001
Synonym N-[2-[[[5-[(Dimethylamino)methyl]furan-2-yl]methyl]sulphanyl]ethyl]-2-nitroacetamide Sodium Salt, Ranitidine Hydrochloride Imp. D (EP), Ranitidine Imp. D (EP), N-[2-[[[5-[(Dimethylamino)methyl]furan-2-yl]-methyl]sulphanyl]ethyl]-2-nitroacetamide Sodium Salt, Ranitidine Hydrochloride Imp. D (EP) as Sodium Salt, AH 24844A, Ph Eur Ranitidine Impurity D, BP Ranitidine Impurity D, Ranitidine complex nitroacetamide hydrochloride salt (AH24844), complex nitroacetamide sodium salt (AH 24844A), Ranitidine Hydrochloride Impurity D as Sodium Salt, Ranitidine Impurity D as Sodium Salt
InChI Formula InChI=1S/C12H19N3O4S.Na/c1-14(2)7-10-3-4-11(19-10)9-20-6-5-13-12(16)8-15(17)18;/h3-4,8,13,16H,5-7,9H2,1-2H3;/q;+1/p-1
SMILES [Na+].CN(C)Cc1oc(CSCCNC(=C[N+](=O)[O-])[O-])cc1
IUPAC Name sodium;1-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethylamino]-2-nitroethenolate
Molecular Weight (g/mol) 323.34
Quantity 25 mg
Formula Weight 323.0916 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.