missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RANGAP1 Recombinant Rabbit Monoclonal Antibody (K01_2P23)

Product Code. 30829614
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829614 50 μL
30824777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829614 Supplier Invitrogen™ Supplier No. MA565646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RANGAP1 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

Ran is a small signaling GTPase that is involved in nucleocytoplasmic transport. Two additional functions of animal Ran in the formation of spindle asters and the reassembly of the nuclear envelope in mitotic cells have been recently reported. In contrast to Ras or Rho, Ran is not associated with membranes. Instead, the spatial sequestering of its accessory proteins, the Ran GTPase-activating protein RanGAP and the nucleotide exchange factor RCC1, appears to define the local concentration of RanGTP vs. RanGDP involved in signaling. Mammalian RanGAP is bound to the nuclear pore by a mechanism involving the attachment of small ubiquitin-related modifier protein (SUMO) to its C terminus and the subsequent binding of the SUMOylated domain to the nucleoporin Nup358.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen RANGAP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2P23
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene RANGAP1
Gene Accession No. P46060, P46061
Gene Alias C79654; fug1; KIAA1835; MGC20266; mKIAA1835; RAN GTPase activating protein 1; RAN GTPase activating protein 1 a; Ran GTPase activating protein 1 b; Ran GTPase activating protein 1 L homeolog; Ran GTPase activating protein 1 S homeolog; ran GTPase-activating protein 1; RANGAP; RanGAP1; rangap1.L; rangap1.S; rangap1a; rangap1-a; rangap1b; rangap1-b; rna1p; SD; segregation distorter homolog; segregation distortion; XELAEV_18023679mg; XELAEV_18025680mg
Gene Symbols RANGAP1
Host Species Rabbit
Immunogen A synthetic peptide of human RanGAP1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19387, 362965, 5905
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.