missing translation for 'onlineSavingsMsg'
Learn More

RanGAP1 Rabbit anti-Human, Mouse, Rat, Clone: 1E7R7, Novus Biologicals™

Product Code. 18362772
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362772 100 μg 100µL
18353503 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18362772 Supplier Bio-Techne Supplier No. NBP316697100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RanGAP1 Monoclonal antibody specifically detects RanGAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RanGAP1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1E7R7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias KIAA1835Fug1, MGC20266, Ran GTPase activating protein 1, ran GTPase-activating protein 1, RanGAP1, SD, segregation distorter homolog, segregation distorter homolog (Drosophila), segregation distortion
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RanGAP1 (P46060).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 5905
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.