missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RANGAP1 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149410
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Ran is a small signaling GTPase that is involved in nucleocytoplasmic transport. Two additional functions of animal Ran in the formation of spindle asters and the reassembly of the nuclear envelope in mitotic cells have been recently reported. In contrast to Ras or Rho, Ran is not associated with membranes. Instead, the spatial sequestering of its accessory proteins, the Ran GTPase-activating protein RanGAP and the nucleotide exchange factor RCC1, appears to define the local concentration of RanGTP vs. RanGDP involved in signaling. Mammalian RanGAP is bound to the nuclear pore by a mechanism involving the attachment of small ubiquitin-related modifier protein (SUMO) to its C terminus and the subsequent binding of the SUMOylated domain to the nucleoporin Nup358.
Specifications
| RANGAP1 | |
| Polyclonal | |
| Unconjugated | |
| RANGAP1 | |
| C79654; fug1; KIAA1835; MGC20266; mKIAA1835; RAN GTPase activating protein 1; RAN GTPase activating protein 1 a; Ran GTPase activating protein 1 b; Ran GTPase activating protein 1 L homeolog; Ran GTPase activating protein 1 S homeolog; ran GTPase-activating protein 1; RANGAP; RanGAP1; rangap1.L; rangap1.S; rangap1a; rangap1-a; rangap1b; rangap1-b; rna1p; SD; segregation distorter homolog; segregation distortion; XELAEV_18023679mg; XELAEV_18025680mg | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 5905 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P46060 | |
| RANGAP1 | |
| E. coli-derived recombinant human RanGAP1, Asn418-Val587. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction