Learn More
RANGAP1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1F3A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67146-100UL
Description
The RANGAP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RANGAP1. This antibody recognizes human, mouse, rat antigen. The RANGAP1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| RANGAP1 | |
| 1F3A4 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| Fug1, KIAA1835, RANGAP1, SD | |
| 100 μL | |
| 5905 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |