missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Reagent Additive 1
Description
1 vial (21mL) of diluted, heat-inactivated normal goat serum. Add 250μL of Reagent Additive 1 per 10mL of 1X detection antibody or reagent diluent.
Specifications
Specifications
| For Use With (Application) | ELISA |
| Quantity | 21 mL |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?