missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RANBP9 Recombinant Rabbit Monoclonal Antibody (JE55-10)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA536208
This item is not returnable.
View return policy
Description
RANBP9 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a protein that binds RAN, a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The protein encoded by this gene has also been shown to interact with several other proteins, including met proto-oncogene, homeodomain interacting protein kinase 2, androgen receptor, and cyclin-dependent kinase 11.
Specifications
| RANBP9 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P69566, Q96S59 | |
| RANBP9 | |
| Recombinant protein within Human RANBP9 aa 90-205. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE55-10 | |
| Unconjugated | |
| RANBP9 | |
| B cell antigen receptor Ig beta associated protein 1; B-cell antigen receptor Ig beta-associated protein 1; BPM 90; BPM L; BPM90; BPML; BPM-L; IBAP 1; Ibap1; IBAP-1; Imp 9; Importin 9; novel centrosomal protein RanBPM; RAN binding protein 9; Ran Binding Protein in the Microtubule organizing center; ran binding protein, centrosomal; Ran BP9; ran-binding protein 9; Ran-binding protein M; RanBP 7; RANBP 9; RanBP7; Ranbp9; RANBPM; RGD1561940 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10048, 364686, 56705 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction