missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANBP9 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17755-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that binds RAN, a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The protein encoded by this gene has also been shown to interact with several other proteins, including met proto-oncogene, homeodomain interacting protein kinase 2, androgen receptor, and cyclin-dependent kinase 11.Specifications
| RANBP9 | |
| Polyclonal | |
| Unconjugated | |
| RANBP9 | |
| BPM L, BPM90, RAN binding protein 9, Ran binding protein M, RanBP7, RANBP9, RANBPM | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10048, 364686, 478728, 56705 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P69566, Q96S59 | |
| Ranbp9 | |
| RANBP9 Fusion Protein Ag12112 | |
| 20 μL | |
| Primary | |
| Rat, Canine, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction