missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RANBP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101810
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RANBP6.
RANBP6 (Ran-binding protein 6) belongs to the importin beta family and may function in nuclear protein import as a nuclear transport receptor.
Specifications
| RANBP6 | |
| Polyclonal | |
| Unconjugated | |
| RANBP6 | |
| C630001B19; RAN binding protein 6; Ran BP6; ran-binding protein 6; Ranbp6; Ran-BP6; Ran-GTP binding protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 240614, 26953, 309326 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60518, Q8BIV3 | |
| RANBP6 | |
| A synthesized peptide derived from human RANBP6(Accession O60518), corresponding to amino acid residues E799-K849. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction