missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RANBP3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13444767
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13444767 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13444767 Supplier Invitrogen™ Supplier No. PA530273

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, Raji. Predicted reactivity: Chimpanzee (100%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Ran binding protein 3 (RanBP3) is a member of Ran GTPase binding proteins that is located at the nuclear pore complex (NPC) and plays an essential role in the transport between cytoplasm and nucleus. RANPB3 is expessed as two spliced forms, RanBP3a and RanBP3b, both of which contains two FXFG motifs, characteristic of a subgroup of nucleoporins in addition to nuclear localization signal in the amino-terminal region (amino acid residues 40-57) and a unique acidic carboxyl terminus which is integral to its switch function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANBP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RANBP3
Gene Accession No. Q9H6Z4
Gene Alias 2610024N24Rik; AA408221; RAN binding protein 3; RANB3; ran-binding protein 3; RAN-binding protein-3; Ranbp3
Gene Symbols RANBP3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RanBP3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8498
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.