Learn More
RanBP2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27606-1-AP-150UL
 View more versions of this product
Product Code. 16875725
  • £349.00 / 150µL
Description
The RanBP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RanBP2. This antibody recognizes human antigen. The RanBP2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RanBP2
Polyclonal
Unconjugated
150 μL
5903
IgG
Western Blot, Immunohistochemistry, ELISA
467 μg/mL