missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RanBP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93338
This item is not returnable.
View return policy
Description
RanBP2 Polyclonal specifically detects RanBP2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| RanBP2 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| ANE1, E3 SUMO-protein ligase RanBP2, EC 5.2.1.8, Nuclear pore complex protein Nup358, nucleoporin 358, Nucleoporin Nup358, NUP358TRP2, P270, RAN binding protein 2, Ran-binding protein 2, RanBP2, transformation-related protein 2,358 kDa nucleoporin, TRP1 | |
| Rabbit | |
| 358 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| P49792 | |
| RANBP2 | |
| The immunogen recognized by this antibody maps to a region between residue 2600 and 2650 of human RAN binding protein 2 using the numbering given in entry NP_006258.3 (GeneID 5903). | |
| Affinity Purified | |
| RUO | |
| 5903 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction