missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RanBP1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79815CL1
This item is not returnable.
View return policy
Description
RanBP1 Polyclonal antibody specifically detects RanBP1 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| RanBP1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| HTF9A, MGC88701, RAN binding protein 1, Ran-binding protein 1, RanBP1, ran-specific GTPase-activating protein | |
| The immunogen recognized by this antibody maps to a region between residue 150 and the C-terminus (residue 201) of human RAN Binding Protein 1 using the numbering given in entry NP_002873.1 (GeneID 5902). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 5902 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction