missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586203
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).
Specifications
| RAN | |
| Polyclonal | |
| Unconjugated | |
| RAN | |
| AAF30287; androgen receptor-associated protein 24; ARA24; ARA25; CG1404; CG1404-PA; CG1404-PB; CG1404-PC; CG1404-PD; Chromosome stability protein 17; CNR1; CST17; Dmel\CG1404; Dmel_CG1404; dran; Genetic suppressor of PRP20-1; GM14354p; gsp1; GTPase Ran; GTPase Ran homolog; GTP-binding nuclear protein GSP1/CNR1; GTP-binding nuclear protein Ran; guanosine triphosphatase Ran; I79_024397; l(1)G0075; L8003.19; LD32416p; LD40852p; member RAS oncogene family; OK/SW-cl.81; OTTHUMP00000166078; OTTHUMP00000166102; Q9VZ23; ran; Ran GTPase; Ran GTPase GSP1; ran GTP-binding protein; RAN, member RAS oncogene family; RAN, member RAS oncogene family L homeolog; ran.L; ran-1; Ran10A; RanGTP; RanGTPase; Ran-PA; Ran-PB; Ran-PC; Ran-PD; RanTC4; Rasl2-8; Ras-like protein TC4; ras-related nuclear protein; tc4; XELAEV_18007741mg; xran; YLR293C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19384, 5901, 84509 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P62826, P62827, P62828 | |
| RAN | |
| Synthetic peptide, corresponding to amino acids 170-200 of Human RAN. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction