missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAN Monoclonal Antibody (8D1H12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548588
This item is not returnable.
View return policy
Description
RAN Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA
Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).
Specifications
| RAN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P62826, P62828 | |
| RAN | |
| Purified recombinant fragment of human RAN (AA: 1-216) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Rat, Monkey | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 8D1H12 | |
| Unconjugated | |
| RAN | |
| AAF30287; androgen receptor-associated protein 24; ARA24; ARA25; CG1404; CG1404-PA; CG1404-PB; CG1404-PC; CG1404-PD; Chromosome stability protein 17; CNR1; CST17; Dmel\CG1404; Dmel_CG1404; dran; Genetic suppressor of PRP20-1; GM14354p; gsp1; GTPase Ran; GTPase Ran homolog; GTP-binding nuclear protein GSP1/CNR1; GTP-binding nuclear protein Ran; guanosine triphosphatase Ran; I79_024397; l(1)G0075; L8003.19; LD32416p; LD40852p; member RAS oncogene family; OK/SW-cl.81; OTTHUMP00000166078; OTTHUMP00000166102; Q9VZ23; ran; Ran GTPase; Ran GTPase GSP1; ran GTP-binding protein; RAN, member RAS oncogene family; RAN, member RAS oncogene family L homeolog; ran.L; ran-1; Ran10A; RanGTP; RanGTPase; Ran-PA; Ran-PB; Ran-PC; Ran-PD; RanTC4; Rasl2-8; Ras-like protein TC4; ras-related nuclear protein; tc4; XELAEV_18007741mg; xran; YLR293C | |
| Mouse | |
| Protein G | |
| RUO | |
| 5901, 84509 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction