missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ran Antibody (JE33-28), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32892
This item is not returnable.
View return policy
Description
Ran Monoclonal antibody specifically detects Ran in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot
Specifications
| Ran | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:500-1:1000 | |
| Androgen receptor-associated protein 24, ARA24Gsp1, GTPase Ran, GTP-binding nuclear protein Ran, guanosine triphosphatase Ran, member RAS oncogene family, OK/SW-cl.81, RAN, member RAS oncogene family, RanGTPase, Ras-like protein TC4, Ras-related nuclear protein, TC4 | |
| Synthetic peptide within Human Ran aa 167-216/ 216. (Uniprot: P62826) | |
| 100 μL | |
| Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 5901 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Western Blot | |
| JE33-28 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction