missing translation for 'onlineSavingsMsg'
Learn More

Ran Antibody (ARAN1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

£175.00

Specifications

Antigen Ran
Clone ARAN1
Concentration 2.5 mg/mL
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500
Applications Western Blot
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18577721
View Documents
Novus Biologicals
NB600-1082-0.025ml
0.025 mL
£175.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ran Monoclonal specifically detects Ran in Human, Mouse, Bovine, Xenopus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

Ran
2.5 mg/mL
Western Blot
Unconjugated
Mouse
Membrane Trafficking and Chaperones, Stem Cell Markers
Androgen receptor-associated protein 24, ARA24Gsp1, GTPase Ran, GTP-binding nuclear protein Ran, guanosine triphosphatase Ran, member RAS oncogene family, OK/SW-cl.81, RAN, member RAS oncogene family, RanGTPase, Ras-like protein TC4, Ras-related nuclear protein, TC4
RAN
IgG2b
Protein A or G purified
Ran (ARAN1)
ARAN1
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500
Monoclonal
Purified
RUO
Human
5901
Denatured recombinant protein (Human).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.