missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RAMP3 Antibody (Medella patent anti-RAMP-3) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27994-100ug
This item is not returnable.
View return policy
Description
RAMP3 Monoclonal antibody specifically detects RAMP3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| RAMP3 | |
| Monoclonal | |
| Unconjugated | |
| O60896 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Medella patent anti-RAMP-3 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| calcitonin receptor-like receptor activity modifying protein 3, Calcitonin-receptor-like receptor activity-modifying protein 3, CRLR activity-modifying protein 3, receptor (calcitonin) activity modifying protein 3, receptor (G protein-coupled) activity modifying protein 3, receptor activity modifying protein 3, receptor activity-modifying protein 3, receptor-activity-modifying protein 3 | |
| RAMP3 | |
| 100 μg | |
| Signal Transduction | |
| 10268 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction