missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAMP1 Recombinant Rabbit Monoclonal Antibody (24GB6910)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557207
This item is not returnable.
View return policy
Description
RAMP1 Recombinant Monoclonal Antibody for Western Blot
The protein encoded by this gene is a member of the RAMP family of single-transmembrane-domain proteins, called receptor (calcitonin) activity modifying proteins (RAMPs). RAMPs are type I transmembrane proteins with an extracellular N terminus and a cytoplasmic C terminus. RAMPs are required to transport calcitonin-receptor-like receptor (CRLR) to the plasma membrane. CRLR, a receptor with seven transmembrane domains, can function as either a calcitonin-gene-related peptide (CGRP) receptor or an adrenomedullin receptor, depending on which members of the RAMP family are expressed. In the presence of this (RAMP1) protein, CRLR functions as a CGRP receptor. The RAMP1 protein is involved in the terminal glycosylation, maturation, and presentation of the CGRP receptor to the cell surface.
Specifications
| RAMP1 | |
| Recombinant Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60894, Q9JJ74, Q9WTJ5 | |
| RAMP1 | |
| A synthesized peptide derived from human RAMP1 (aa 139-148). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB6910 | |
| Unconjugated | |
| RAMP1 | |
| 9130218E19Rik; calcitonin receptor-like receptor activity modifying protein 1; calcitonin-receptor-like receptor activity-modifying protein 1; CRLR activity-modifying protein 1; MGC38240; RAMP1; receptor (calcitonin) activity modifying protein 1; receptor (G protein-coupled) activity modifying protein 1; receptor activity modifying protein 1; receptor activity-modifying protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10267, 51801, 58965 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction