missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAMP1, Mouse anti-Human, Clone: 1F1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010267-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: CQEANYGALLRELCLTQFQVDMEAVGETLWCDWGRTIRSYRELADCTWHMAEKLGCFWPNAEVDRFFLAVHGRYFRSCPISGRAVRDPPGSSpecifications
| RAMP1 | |
| Monoclonal | |
| Unconjugated | |
| receptor (G protein-coupled) activity modifying protein 1 | |
| - | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1F1 | |
| In 1x PBS, pH 7.4 | |
| NM_005855 | |
| RAMP1 | |
| RAMP1 (NP_005846, 27 a.a. ∼ 117 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10267 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction