missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAMP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£164.00 - £366.00
Specifications
| Antigen | RAMP1 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18699241
|
Novus Biologicals
NBP2-94078-0.02ml |
0.02 mL |
£164.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681222
|
Novus Biologicals
NBP2-94078-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAMP1 Polyclonal antibody specifically detects RAMP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| RAMP1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 10267 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| calcitonin receptor-like receptor activity modifying protein 1, Calcitonin-receptor-like receptor activity-modifying protein 1, CRLR activity-modifying protein 1, receptor (calcitonin) activity modifying protein 1, receptor (G protein-coupled) activity modifying protein 1, receptor activity modifying protein 1, receptor activity-modifying protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 21-120 of human RAMP1 (NP_005846.1). CPISGRAVRDPPGS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title