missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Binder™ Ramp for Environmental Walk-in Chambers
Shop All BINDER Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Description
- Accessory for all WIC models
Specifications
Specifications
| Color | Gray |
| For Use With (Equipment) | Binder™ Environmental Walk-in Chamber |
| Material | Stainless Steel |
| Type | WIC Ramp |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction