missing translation for 'onlineSavingsMsg'
Learn More

Ramosetron Hydrochloride, MedChemExpress

Product Code. 16484407
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 16484407

Brand: Medchem Express HYB0595

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ramosetron Hydrochloride(YM060 Hydrochloride) is a serotonin 5-HT3 receptor antagonist for the treatment of nausea and vomiting.

Ramosetron Hydrochloride(YM060 Hydrochloride) is a serotonin 5-HT3 receptor antagonist for the treatment of nausea and vomiting.

Specifications

Chemical Name or Material Ramosetron Hydrochloride
CAS 132907-72-3
Color White
Purity Grade Notes Research
Molecular Formula C17H18ClN3O
Quantity 50 mg
Synonym YM060
Solubility Information H2O : 100 mg/mL (316.66 mM; Need ultrasonic) ∣DMSO : 32 mg/mL (101.33 mM; Need ultrasonic and warming)
SMILES O=C(C1=CN(C)C2=C1C=CC=C2)[C@@H](C3)CCC4=C3NC=N4.Cl
Molecular Weight (g/mol) 315.8
Formula Weight 315.8
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.