missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ramipril

Product Code. 18704719
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18704719

Brand: Tocris Bioscience™ 5250/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Non-peptide, competitive angiotensin-converting enzyme (ACE) inhibitor

Ramipril is a non-peptide, competitive angiotensin converting enzyme (ACE) inhibitor (IC50 = 5 nM). Prodrug which is hydrolyzed in vivo to the active metabolite ramiprilat. Displays protective effects on endothelium against high-glucose induced dysfunction. Exhibits antihypertensive effects.
TRUSTED_SUSTAINABILITY

Specifications

Purity >99%
Quantity 50 mg
Product Type Ramipril
Molecular Weight (g/mol) 416.51
Inhibitors Ramipril

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.