missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ramipril Methyl Ester(>85%), TRC
Specifications
Specifications
| Chemical Name or Material | Ramipril methyl ester |
| CAS | 108313-11-7 |
| Molecular Formula | C22 H30 N2 O5 |
| Quantity | 100 mg |
| Synonym | (2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-(Methoxycarbonyl)-3-phenylpropyl]amino]propanoyl]octahydrocyclopenta[b]pyrrole-2-carboxylic Acid, Ramipril Methyl Ester, Ramipril Imp. A (EP), Ramipril Related Compound A (USP) |
| InChI Formula | InChI=1S/C22H30N2O5/c1-14(20(25)24-18-10-6-9-16(18)13-19(24)21(26)27)23-17(22(28)29-2)12-11-15-7-4-3-5-8-15/h3-5,7-8,14,16-19,23H,6,9-13H2,1-2H3,(H,26,27)/t14-,16-,17-,18-,19-/m0/s1 |
| SMILES | COC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2[C@H]3CCC[C@H]3C[C@H]2C(=O)O |
| IUPAC Name | (2S,3aS,6aS)-1-[(2S)-2-[[(2S)-1-methoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]pyrrole-2-carboxylic acid |
| Molecular Weight (g/mol) | 402.48 |
| Formula Weight | 402.2155 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?