missing translation for 'onlineSavingsMsg'
Learn More

Ramipril, MedChemExpress

Product Code. 16498206
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498206 100 mg 100mg
16432086 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498206 Supplier Medchem Express Supplier No. HYB0279100mg
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ramipril (HOE-498) is an angiotensin-converting enzyme (ACE) inhibitor with IC50 of 5 nM.

Ramipril (HOE-498) is an angiotensin-converting enzyme (ACE) inhibitor with IC50 of 5 nM.

Specifications

Chemical Name or Material Ramipril
CAS 87333-19-5
Color White
Purity Grade Notes Research
Molecular Formula C23H32N2O5
Quantity 100 mg
Synonym HOE-498
Solubility Information DMSO : ≥ 100 mg/mL (240.09 mM) ∣H2O : 1 mg/mL (2.40 mM; Need ultrasonic)
SMILES O=C([C@@H]1C[C@@](CCC2)([H])[C@@]2([H])N1C([C@@H](N[C@H](C(OCC)=O)CCC3=CC=CC=C3)C)=O)O
Molecular Weight (g/mol) 416.51
Formula Weight 416.51
Percent Purity 98.16%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.