missing translation for 'onlineSavingsMsg'
Learn More

Ramelteon, MedChemExpress

Product Code. 16498258
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498258 10 mg 10mg
16484147 50 mg 50mg
16467986 100 mg 100mg
16411976 500 mg 500mg
16410708 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498258 Supplier Medchem Express Supplier No. HYA0014
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ramelteon is a potent, highly selective, and orally active agonist of MT1/MT2 with Ki values of 14 and 112 pM, respectively. Ramelteon has the potential for the research of insomnia. Ramelteon consistently reduces sleep onset after long-term treatment, with no next-morning residual effects or rebound insomnia or withdrawal symptoms upon discontinuation.

Ramelteon is a potent, highly selective, and orally active agonist of MT1/MT2 with Ki values of 14 and 112 pM, respectively. Ramelteon has the potential for the research of insomnia. Ramelteon consistently reduces sleep onset after long-term treatment, with no next-morning residual effects or rebound insomnia or withdrawal symptoms upon discontinuation.

Specifications

Chemical Name or Material Ramelteon
CAS 196597-26-9
Color White
Purity Grade Notes Research
Molecular Formula C16H21NO2
Quantity 10 mg
Synonym TAK-375
Solubility Information DMSO : ≥ 50 mg/mL (192.80 mM)
SMILES O=C(NCC[C@H]1C2=C(C=CC3=C2CCO3)CC1)CC
Molecular Weight (g/mol) 259.34
Formula Weight 259.34
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.