missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALYL Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17179-1-AP-150UL
This item is not returnable.
View return policy
Description
RALYL belongs to the RRM HNRPC family, RALY subfamily. It contains 1 RRM (RNA recognition motif) domain. The functions of RALYL remain unknown.Specifications
| RALYL | |
| Polyclonal | |
| Unconjugated | |
| RALYL | |
| hnRNP core protein C like 3, HNRPCL3, hRALYL, RALY RNA binding protein like, RALYL, RNA binding Raly like protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 138046, 76897 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86SE5, Q8BTF8 | |
| Ralyl | |
| RALYL Fusion Protein Ag10898 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction