missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RALYL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566235
This item is not returnable.
View return policy
Description
Immunogen sequence: DYHGRVPPPPR AVIPLKRPRV AVTTTRRGKG VFSMKGGSRS TASGSTGSKL KSD Highest antigen sequence identity to the following orthologs - mouse 91%, rat 89%.
RALYL belongs to the RRM HNRPC family, RALY subfamily. It contains 1 RRM (RNA recognition motif) domain. The functions of RALYL remain unknown.
Specifications
| RALYL | |
| Polyclonal | |
| Unconjugated | |
| RALYL | |
| 0710005M24Rik; Heterogeneous nuclear ribonucleoprotein C-like 3; hnRNP core protein C-like 3; HNRPCL3; hRALYL; RALY RNA binding protein like; RALY RNA binding protein-like; RALYL; RNA-binding Raly-like protein; Unknown (protein for MGC:133552) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 138046 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86SE5 | |
| RALYL | |
| Recombinant Human RALYL. Recombinant protein control fragment (Product #RP-106917). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction