Learn More
RALYL Mouse anti-Human, Unconjugated, Clone: 1F3F4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67534-1-IG-20UL
 View more versions of this product
Product Code. 16880216
  • £135.00 / 20µL
Description
The RALYL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RALYL. This antibody recognizes human, mouse, rat, pig antigen. The RALYL antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
RALYL
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunofluorescence, ELISA
1F3F4
Unconjugated
20 μL
138046
IgG1