Learn More
RALYL Mouse anti-Human, Unconjugated, Clone: 1F3F4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67534-1-IG-150UL
Description
The RALYL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RALYL. This antibody recognizes human, mouse, rat, pig antigen. The RALYL antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| RALYL | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunofluorescence, ELISA | |
| 1F3F4 | |
| Unconjugated | |
| 150 μL | |
| 138046 | |
| IgG1 |