missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RALY Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583671
This item is not returnable.
View return policy
Description
Immunogen sequence: RLFDYRGRLS PVPVPRAVPV KRPRVTVPLV RRVKTNVPVK LFARSTAVTT SSAKIKLKSS ELQAIK Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 94%.
In infectious mononucleosis, anti-EBNA-1 antibodies are produced which cross-react with multiple normal human proteins. The cross-reactivity is due to anti-gly/ala antibodies that cross-react with host proteins containing configurations like those in the EBNA-1 repeat. One such antigen is RALY which is a member of the heterogeneous nuclear ribonucleoprotein gene family.
Specifications
| RALY | |
| Polyclonal | |
| Unconjugated | |
| RALY | |
| AI663842; Autoantigen p542; autoantigenic hnRNP-associated with lethal yellow; heterogeneous nuclear ribonucleoprotein C-like 2; hnR; hnRNP associated with lethal yellow protein; hnRNP associated with lethal yellow protein homolog; hnRNP core protein C-like 2; hnRNP-associated with lethal yellow; HNRPCL2; LOW QUALITY PROTEIN: RNA-binding protein Raly; maternally-expressed hnRNP C-related protein; Merc; MGC117312; P542; RALY; RALY heterogeneous nuclear ribonucleoprotein; RALY protein; RNA binding protein; RNA binding protein (autoantigenic, hnRNP-associated with lethal yellow) short isoform; RNA binding protein, autoantigenic (hnRNP-associated with lethal yellow homolog (mouse)); RNA binding protein, autoantigenic (hnRNP-associated with lethal yellow homolog); RNA-binding protein (autoantigenic); RNA-binding protein (autoantigenic, hnRNP-associated with lethal yellow); RNA-binding protein Raly; zgc:103445 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19383, 22913, 296301 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q64012, Q9UKM9 | |
| RALY | |
| Recombinant protein corresponding to Human RALY. Recombinant protein control fragment (Product #RP-99423). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction