missing translation for 'onlineSavingsMsg'
Learn More

Raltitrexed, MedChemExpress

Product Code. p-200101061
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16495136

Brand: Medchem Express HY10821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Raltitrexed is an antimetabolite drug used in chemotherapy, acting by inhibiting thymidylate synthase.

Raltitrexed is an antimetabolite drug used in chemotherapy, acting by inhibiting thymidylate synthase.

Specifications

Chemical Name or Material Raltitrexed
Physical Form Powder
CAS 112887-68-0
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H22N4O6S
For Use With (Application) Cancer-programmed cell death
Synonym ZD1694 D1694 ICI-D1694
Solubility Information DMSO : ≥ 29 mg/mL (63.25 mM)
SMILES CC(NC1=O)=NC2=C1C=C(CN(C)C3=CC=C(C(N[C@H](C(O)=O)CCC(O)=O)=O)S3)C=C2
Molecular Weight (g/mol) 458.49
Formula Weight 458.49
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.