missing translation for 'onlineSavingsMsg'
Learn More

Raltitrexed, MedChemExpress

Product Code. 16495136
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495136 100 mg 100mg
16480527 50 mg 50mg
16474178 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16495136 Supplier Medchem Express Supplier No. HY10821
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Raltitrexed is an antimetabolite drug used in chemotherapy, acting by inhibiting thymidylate synthase.

Raltitrexed is an antimetabolite drug used in chemotherapy, acting by inhibiting thymidylate synthase.

Specifications

Chemical Name or Material Raltitrexed
Physical Form Powder
CAS 112887-68-0
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H22N4O6S
For Use With (Application) Cancer-programmed cell death
Synonym ZD1694 D1694 ICI-D1694
Solubility Information DMSO : ≥ 29 mg/mL (63.25 mM)
SMILES CC(NC1=O)=NC2=C1C=C(CN(C)C3=CC=C(C(N[C@H](C(O)=O)CCC(O)=O)=O)S3)C=C2
Molecular Weight (g/mol) 458.49
Formula Weight 458.49
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.