missing translation for 'onlineSavingsMsg'
Learn More

Raloxifene hydrochloride, MedChemExpress

Product Code. 16421987
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421987 50 mg 50mg
16416306 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16421987 Supplier Medchem Express Supplier No. HY13738A
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Raloxifene hydrochloride (Keoxifene hydrochloride) is a second generation selective and orally active estrogen receptor modulator. Raloxifene hydrochloride produces estrogen-agonistic effects on bone and lipid metabolism and estrogen-antagonistic effects on uterine endometrium and breast tissue.

Raloxifene hydrochloride (Keoxifene hydrochloride) is a second generation selective and orally active estrogen receptor modulator. Raloxifene hydrochloride produces estrogen-agonistic effects on bone and lipid metabolism and estrogen-antagonistic effects on uterine endometrium and breast tissue.

Specifications

Chemical Name or Material Raloxifene hydrochloride
CAS 82640-04-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H28ClNO4S
Quantity 50 mg
Synonym Keoxifene hydrochloride LY156758LY139481 hydrochloride
Solubility Information DMSO : 33.33 mg/mL (65.35 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.96 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1=C(C2=CC=C(O)C=C2)SC3=CC(O)=CC=C31)C4=CC=C(OCCN5CCCCC5)C=C4.Cl
Molecular Weight (g/mol) 510.04
Formula Weight 510.04
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.