missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Raloxifene hydrochloride

Product Code. 17211242 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17211242 5 g 5g
17201242 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17211242 Supplier Thermo Scientific Chemicals Supplier No. 466500050
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82640-04-8 | C28H28ClNO4S, C28H27NO4S·HCl | 510.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82640-04-8
Molecular Formula C28H27NO4S·HCl

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: .. Other solubilities: soluble in dmso
Melting Point 263.0°C
Formula Weight 510.04
Chemical Name or Material Raloxifene hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.