missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Raloxifene (Hydrochloride)
Description
- Raloxifene is a selective estrogen receptor modulator (SERM) that exhibits agonistic (estrogenic) activity in bone cells without stimulating ER activity in breast or uterine tissues (Black et al.)
- Enhances osteoblast differentiation in mouse bone marrow and human osteoblast cultures (Qu et al.; Viereck et al.)
- Inhibits osteoclast differentiation in primary human bone marrow mononuclear cell cultures (Ramalho et al.)
- Reduced endodermal differentiation (HNF-4α expression) in embryoid bodies derived from human embryonic stem (ES) cells (Kim et al.)
Specifications
Specifications
| CAS | 82640-04-8 |
| Synonym | Keoxifene; LY156758 |
| Quantity | 100 mg |
| Molecular Formula | C28H27NO4S · HCl |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?