missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALGPS1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93901-0.02ml
This item is not returnable.
View return policy
Description
RALGPS1 Polyclonal antibody specifically detects RALGPS1 in Rat samples. It is validated for Western Blot
Specifications
| RALGPS1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| KIAA0351RalGEF 2, Ral GEF with PH domain and SH3 binding motif 1, Ral GEF with PH domain and SH3-binding motif 1, Ral guanine nucleotide exchange factor 2, Ral guanine nucleotide exchange factor RalGPS1A, RalA exchange factor RalGPS1, RALGEF2RALGPS1A, ras-specific guanine nucleotide-releasing factor RalGPS1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RALGPS1 (NP_055451.1). MYKRNGLMASVLVTSATPQGSSSSDSLEGQSCDYASKSYDAVVFDVLKVTPEEFASQITLMDIPVFKAIQPEELASCGWSKKEKHSLAPNVVAFTRRFNQVSFWVVREILTAQTLKIRAEILSHFVKIAKKLLELNNLHSLMSVVSALQSAPIFRLTKTWALLNRKDKTTFEKLDYLMSKEDNYKRTREYIRSLKMVPSIPYLGIYLLDLIYIDSAYPASGSIMENEQRSNQMNNILRIIADLQVSCSYDHLTTLPHVQKYLKSVRYIEELQKFVEDDNYKLSLRIEPGSSSPRLVSSKE | |
| 0.02 mL | |
| Signal Transduction | |
| 9649 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction