missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RALGAPA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101287
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total AS250.
Catalytic subunit of the heterodimeric RalGAP2 complex which acts as a GTPase activator for the Ras-like small GTPases RALA and RALB.
Specifications
| RALGAPA2 | |
| Polyclonal | |
| Unconjugated | |
| RALGAPA2 | |
| 250 kDa substrate of Akt; A230067G21Rik; akt substrate AS250; AS250; bA287B20.1; BC053994; C20orf74; dJ1049G11; dJ1049G11.4; Kiaa1272; p220; pp250; Ral GTPase activating protein catalytic alpha subunit 2; Ral GTPase activating protein, alpha subunit 2 (catalytic); ral GTPase-activating protein alpha subunit 2; ral GTPase-activating protein subunit alpha-2; RALGAPA2; RapGAPalpha2; RGC2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 241694, 57186 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A3KGS3, Q2PPJ7 | |
| RALGAPA2 | |
| A synthesized peptide derived from human RALGAPA2(Accession Q2PPJ7), corresponding to amino acid residues S646-S696. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction