missing translation for 'onlineSavingsMsg'
Learn More

Ralfinamide, MedChemExpress

Product Code. 16429896
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429896 50 mg 50mg
16423368 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16429896 Supplier Medchem Express Supplier No. HY101437

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ralfinamide (FCE-26742A) is an orally available Na+ blocker derived from α-aminoamide, with function of suppressing pain.

Ralfinamide (FCE-26742A) is an orally available Na+ blocker derived from α-aminoamide, with function of suppressing pain.

Specifications

Chemical Name or Material Ralfinamide
CAS 133865-88-0
Color White
Purity Grade Notes Research
Molecular Formula C17H19FN2O2
Quantity 50 mg
Synonym FCE-26742A
Solubility Information DMSO : ≥ 125 mg/mL (413.44 mM)
SMILES C[C@H](NCC1=CC=C(OCC2=CC=CC=C2F)C=C1)C(N)=O
Molecular Weight (g/mol) 302.34
Formula Weight 302.34
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.