missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALBP1 Antibody (OTI11B2), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73788PCP
This item is not returnable.
View return policy
Description
RALBP1 Monoclonal specifically detects RALBP1 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.
Specifications
| RALBP1 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| RALBP1 | |
| Full length recombinant protein of human RALBP1 (NP_006779) produced in HEK293T cells. | |
| 0.1 mL | |
| 10928.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI11B2 | |
| Flow Cytometry | |
| Dinitrophenyl S-glutathione ATPase, ralA binding protein 1, ralA-binding protein 1,76 kDa Ral-interacting protein, RalBP1, Ral-interacting protein 1, RIP, RIP1, RLIP1, RLIP76DNP-SG ATPase | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction