Learn More
RALB Mouse anti-Human, CoraLite 488, Clone: 1D11C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67094-100UL
Description
The RALB antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human RALB. This antibody recognizes human, rat, pig, mouse antigen. The RALB antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| RALB | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 5899 | |
| IgG2a |
| Immunofluorescence | |
| 1D11C11 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |