missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RALA Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584684
This item is not returnable.
View return policy
Description
Immunogen sequence: VEDYEPTKAD SYRKKVVLDG EEVQID Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
Multifunctional GTPase involved in a variety of cellular processes including gene expression, cell migration, cell proliferation, oncogenic transformation and membrane trafficking. Accomplishes its multiple functions by interacting with distinct downstream effectors. Acts as a GTP sensor for GTP-dependent exocytosis of dense core vesicles. Plays a role in the early stages of cytokinesis and is required to tether the exocyst to the cytokinetic furrow. The RALA-exocyst complex regulates integrin-dependent membrane raft exocytosis and growth signaling. Key regulator of LPAR1 signaling and competes with GRK2 for binding to LPAR1 thus affecting the signaling properties of the receptor. Required for anchorage-independent proliferation of transformed cells.
Specifications
| RALA | |
| Polyclonal | |
| Unconjugated | |
| Rala | |
| 3010001O15Rik; AW322615; MGC48949; Ral; -ral simian leukemia viral oncogene homolog A (ras related); RALA; Ral-a; ral-A protein; RALA Ras like proto-oncogene A; Ras family small GTP binding protein RALA; ras related GTP binding protein A; Rasl1; RAS-like protein A; RAS-like, family 1; ras-related protein Ral-A; v-ral simian leukemia viral oncogene homolog A (ras related) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5898 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P11233 | |
| Rala | |
| Recombinant protein corresponding to Human RALA. Recombinant protein control fragment (Product #RP-104540). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction