missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAIDD Recombinant Rabbit Monoclonal Antibody (3Z9N8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538173
This item is not returnable.
View return policy
Description
Positive Samples: HT-29, 293T, K-562 Immunogen sequence: GDRLTGIPSH ILNSSPSDRQ INQLAQRLGP EWEPMVLSLG LSQTDIYRCK ANHPHNVQSQ VVEAFIRWRQ RFGKQATFQS LHNGLRAVEV DPSLLLHMLE.
The protein encoded by this gene is a death domain (CARD/DD)-containing protein and has been shown to induce cell apoptosis. Through its CARD domain, this protein interacts with, and thus recruits, caspase 2/ICH1 to the cell death signal transduction complex that includes tumor necrosis factor receptor 1 (TNFR1A), RIPK1/RIP kinase, and numbers of other CARD domain-containing proteins.
Specifications
| RAIDD | |
| Recombinant Monoclonal | |
| 0.65 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P78560 | |
| CRADD | |
| A synthetic peptide corresponding to a sequence within amino acids 100-199 of human RAIDD/CRADD (NP_0037961). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 3Z9N8 | |
| Unconjugated | |
| CRADD | |
| CASP2 and RIPK1 domain containing adaptor with death domain; CASP2 and RIPK1 domain-containing adaptor with death domain; caspase and RIP adapter with death domain; caspase and RIP adaptor with death domain; CRADD; death adaptor molecule RAIDD; death domain containing protein CRADD; death domain-containing protein CRADD; MGC9163; MRT34; RAIDD; RIP-associated ICH1/CED3-homologous protein with death domain; RIP-associated protein with a death domain | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 314756, 8738 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction