missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAIDD Polyclonal Antibody
GREENER_CHOICE

Product Code. 12838082
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12838082 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12838082 Supplier Invitrogen™ Supplier No. PA519865

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~22kDa. A suggested positive control is Hela cell lysate.

The protein encoded by this gene is a death domain (CARD/DD)-containing protein and has been shown to induce cell apoptosis. Through its CARD domain, this protein interacts with, and thus recruits, caspase 2/ICH1 to the cell death signal transduction complex that includes tumor necrosis factor receptor 1 (TNFR1A), RIPK1/RIP kinase, and numbers of other CARD domain-containing proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAIDD
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CRADD
Gene Accession No. P78560
Gene Alias CASP2 and RIPK1 domain containing adaptor with death domain; CASP2 and RIPK1 domain-containing adaptor with death domain; caspase and RIP adapter with death domain; caspase and RIP adaptor with death domain; CRADD; death adaptor molecule RAIDD; death domain containing protein CRADD; death domain-containing protein CRADD; MGC9163; MRT34; RAIDD; RIP-associated ICH1/CED3-homologous protein with death domain; RIP-associated protein with a death domain
Gene Symbols CRADD
Host Species Rabbit
Immunogen A peptide corresponding to amino acids near the carboxy terminus of human RAIDD.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8738
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.