missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAIDD Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148883
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a death domain (CARD/DD)-containing protein and has been shown to induce cell apoptosis. Through its CARD domain, this protein interacts with, and thus recruits, caspase 2/ICH1 to the cell death signal transduction complex that includes tumor necrosis factor receptor 1 (TNFR1A), RIPK1/RIP kinase, and numbers of other CARD domain-containing proteins.
Specifications
| RAIDD | |
| Polyclonal | |
| Unconjugated | |
| CRADD | |
| CASP2 and RIPK1 domain containing adaptor with death domain; CASP2 and RIPK1 domain-containing adaptor with death domain; caspase and RIP adapter with death domain; caspase and RIP adaptor with death domain; CRADD; death adaptor molecule RAIDD; death domain containing protein CRADD; death domain-containing protein CRADD; MGC9163; MRT34; RAIDD; RIP-associated ICH1/CED3-homologous protein with death domain; RIP-associated protein with a death domain | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 12905, 314756, 8738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O88843, P78560 | |
| CRADD | |
| E. coli-derived recombinant mouse CRADD, Met1-Glu199. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction