missing translation for 'onlineSavingsMsg'
Learn More

RAIDD/CRADD Antibody, DyLight 680, Novus Biologicals™

Product Code. 18335965 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335965 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335965 Supplier Novus Biologicals Supplier No. NBP300051FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAIDD/CRADD Polyclonal specifically detects RAIDD/CRADD in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAIDD/CRADD
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias CASP2 and RIPK1 domain containing adaptor with death domain, Caspase and RIP adapter with death domain, caspase and RIP adaptor with death domain, death domain containing protein CRADD, death domain-containing protein CRADD, MGC9163, RAIDDdeath adaptor molecule RAIDD, RIP-associated ICH1/CED3-homologous protein with death domain, RIP-associated protein with a death domain
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant RAIDD/CRADD (P78560; Met1-Glu199).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8738
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.