missing translation for 'onlineSavingsMsg'
Learn More

RAI3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15362611
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15362611 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15362611 Supplier Invitrogen Supplier No. PA533796

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RAI3 Polyclonal Antibody

RAI3 Polyclonal Antibody for ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAI3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPRC5A
Gene Accession No. Q8NFJ5
Gene Alias G protein-coupled receptor class C group 5 member A; G protein-coupled receptor, class C, group 5, member A; G protein-coupled receptor, family C, group 5, member A; GPCR5A; Gprc5a; G-protein coupled receptor family C group 5 member A; orphan G-protein-coupling receptor PEIG-1; PEIG-1; Phorbol ester induced gene 1; phorbol ester induced protein-1; Rai3; Raig1; RAIG-1; retinoic acid induced 3; retinoic acid responsive; retinoic acid-induced gene 1 protein; Retinoic acid-induced protein 3; TIG1; TPA induced gene 1
Gene Symbols GPRC5A
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human GPRC5A / RAI3.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9052
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.